Assay Details
| Gene Symbol: TPM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003432905.2 | XP_003432953.1 | 6 - 7 | 776 | 118 | |
| XM_003432906.4 | XP_003432954.1 | 6 - 7 | 715 | 118 | ||
| XM_847548.5 | XP_852641.1 | 7 - 8 | 761 | 118 | ||
| XM_861034.5 | XP_866127.1 | 7 - 8 | 767 | 118 | ||
| GenBank mRNA | KR822489.1 | - | 7 - 8 | 703 | 118 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TPM4
Genomic Map