Assay Details
| Gene Symbol: CENPQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022426136.1 | XP_022281844.1 | 4 - 5 | 411 | 140 | |
| XM_022426137.1 | XP_022281845.1 | 4 - 5 | 461 | 140 | ||
| XM_022426138.1 | XP_022281846.1 | 4 - 5 | 426 | 140 | ||
| XM_022426139.1 | XP_022281847.1 | 4 - 5 | 444 | 140 | ||
| XM_022426140.1 | XP_022281848.1 | 4 - 5 | 357 | 140 | ||
More Information
Set Membership: |
|

CENPQ
Genomic Map