Assay Details
| Gene Symbol: TCP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627248.3 | XP_005627305.1 | 3 - 4 | 428 | 69 | |
| XM_022425962.1 | XP_022281670.1 | 3 - 4 | 330 | 69 | ||
| XM_022425963.1 | XP_022281671.1 | 3 - 4 | 239 | 69 | ||
| XM_532117.6 | XP_532117.4 | 3 - 4 | 418 | 69 | ||
More Information
Set Membership: |
|

TCP11
Genomic Map