Assay Details
| Gene Symbol: PRIMPOL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629990.2 | XP_005630047.1 | 8 - 9 | 1256 | 137 | |
| XM_005629991.3 | XP_005630048.1 | 8 - 9 | 1341 | 137 | ||
| XM_005629992.3 | XP_005630049.1 | 8 - 9 | 1344 | 137 | ||
| XM_014120170.2 | XP_013975645.1 | 8 - 9 | 1343 | 137 | ||
| XM_532846.6 | XP_532846.3 | 8 - 9 | 1266 | 137 | ||
More Information
Set Membership: |
|

PRIMPOL
Genomic Map