Assay Details
| Gene Symbol: LOC612942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022425650.1 | XP_022281358.1 | 2 - 3 | 557 | 71 | |
| XM_022425651.1 | XP_022281359.1 | 3 - 4 | 668 | 71 | ||
| XM_022425652.1 | XP_022281360.1 | 3 - 4 | 673 | 71 | ||
More Information
Set Membership: |
|

LOC612942
Genomic Map