Assay Details
| Gene Symbol: LOC612942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022425650.1 | XP_022281358.1 | 4 - 5 | 714 | 123 | |
| XM_022425651.1 | XP_022281359.1 | 5 - 6 | 825 | 123 | ||
| XM_022425652.1 | XP_022281360.1 | 5 - 6 | 830 | 123 | ||
More Information
Set Membership: |
|

LOC612942
Genomic Map