Assay Details
| Gene Symbol: C12H6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_532058.6 | XP_532058.3 | 4 - 5 | 956 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C12H6orf136| Gene Symbol: C12H6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_532058.6 | XP_532058.3 | 4 - 5 | 956 | 74 | |
Set Membership: |
|