Assay Details
| Gene Symbol: BICRAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627362.3 | XP_005627419.1 | 7 - 8 | 2165 | 86 | |
| XM_005627363.3 | XP_005627420.1 | 7 - 8 | 2149 | 86 | ||
| XM_014118453.2 | XP_013973928.1 | 7 - 8 | 2482 | 86 | ||
More Information
Set Membership: |
|

BICRAL
Genomic Map