Assay Details
| Gene Symbol: BICRAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627362.3 | XP_005627419.1 | 9 - 10 | 2375 | 59 | |
| XM_005627363.3 | XP_005627420.1 | 9 - 10 | 2359 | 59 | ||
| XM_014118453.2 | XP_013973928.1 | 9 - 10 | 2692 | 59 | ||
More Information
Set Membership: |
|

BICRAL
Genomic Map