Assay Details
| Gene Symbol: MRPS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627350.3 | XP_005627407.1 | 1 - 2 | 113 | 81 | |
| XM_532137.6 | XP_532137.2 | 1 - 2 | 113 | 81 | ||
More Information
Set Membership: |
|

MRPS10
Genomic Map