Assay Details
| Gene Symbol: GLRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629375.3 | XP_005629432.1 | 7 - 8 | 844 | 55 | |
| XM_532701.7 | XP_532701.1 | 8 - 9 | 1269 | 55 | ||
More Information
Set Membership: |
|

GLRB
Genomic Map