Assay Details
| Gene Symbol: CD59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022405336.1 | XP_022261044.1 | 2 - 3 | 104 | 73 | |
| XM_533156.5 | XP_533156.1 | 2 - 3 | 164 | 73 | ||
More Information
Set Membership: |
|

CD59
Genomic Map