Assay Details
| Gene Symbol: COX7A2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022404673.1 | XP_022260381.1 | 2 - 3 | 152 | 120 | |
| XM_532946.6 | XP_532946.1 | 2 - 3 | 289 | 120 | ||
More Information
Set Membership: |
|

COX7A2L
Genomic Map