Assay Details
| Gene Symbol: CHST10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626011.2 | XP_005626068.1 | 5 - 6 | 535 | 90 | |
| XM_005626012.2 | XP_005626069.1 | 4 - 5 | 450 | 90 | ||
| XM_005626013.2 | XP_005626070.1 | 2 - 3 | 224 | 90 | ||
More Information
Set Membership: |
|

CHST10
Genomic Map