Assay Details
| Gene Symbol: CYCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197045.1 | NP_001183974.1 | 1 - 2 | 121 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001197045.1 transcripts.
Set Membership: |
|

CYCS
Genomic Map