Assay Details
| Gene Symbol: WASHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117392.2 | XP_013972867.1 | 20 - 21 | 2147 | 138 | |
| XM_531762.6 | XP_531762.3 | 21 - 22 | 2264 | 138 | ||
More Information
Set Membership: |
|

WASHC4
Genomic Map