Assay Details
| Gene Symbol: GARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639490.4 | XP_003639538.1 | 3 - 4 | 451 | 90 | |
| XM_014119146.2 | XP_013974621.1 | 3 - 4 | 273 | 90 | ||
More Information
Set Membership: |
|

GARS1
Genomic Map