Assay Details
| Gene Symbol: GPATCH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630336.3 | XP_005630393.1 | 8 - 9 | 920 | 75 | |
| XM_532830.6 | XP_532830.3 | 8 - 9 | 904 | 75 | ||
More Information
Set Membership: |
|

GPATCH11
Genomic Map