Assay Details
Gene Symbol: C12H6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627281.2 | XP_005627338.1 | 5 - 6 | 820 | 79 | |
XM_005627282.2 | XP_005627339.1 | 6 - 7 | 980 | 79 | ||
XM_005627283.3 | XP_005627340.1 | 4 - 5 | 782 | 79 | ||
XM_014118402.2 | XP_013973877.1 | 3 - 4 | 453 | 79 | ||
XM_022425987.1 | XP_022281695.1 | 6 - 7 | 880 | 79 | ||
XM_022425988.1 | XP_022281696.1 | 5 - 6 | 787 | 79 | ||
XM_022425989.1 | XP_022281697.1 | 5 - 6 | 878 | 79 | ||
XM_532127.5 | XP_532127.2 | 4 - 5 | 730 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
