Assay Details
| Gene Symbol: ZNF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628113.3 | XP_005628170.1 | 2 - 3 | 159 | 91 | |
| XM_014118757.2 | XP_013974232.1 | 2 - 3 | 651 | 91 | ||
| XM_014118758.2 | XP_013974233.1 | 2 - 3 | 644 | 91 | ||
| XM_022426892.1 | XP_022282600.1 | 2 - 3 | 653 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005628113.3 transcripts.
Set Membership: |
|

ZNF7
Genomic Map