Assay Details
| Gene Symbol: ZNF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628110.3 | XP_005628167.1 | 3 - 4 | 766 | 73 | |
| XM_005628111.3 | XP_005628168.1 | 4 - 5 | 431 | 73 | ||
| XM_005628113.3 | XP_005628170.1 | 4 - 5 | 397 | 73 | ||
| XM_014118754.2 | XP_013974229.1 | 5 - 6 | 561 | 73 | ||
| XM_014118756.2 | XP_013974231.1 | 4 - 5 | 922 | 73 | ||
| XM_014118757.2 | XP_013974232.1 | 4 - 5 | 889 | 73 | ||
| XM_014118758.2 | XP_013974233.1 | 4 - 5 | 882 | 73 | ||
| XM_022426891.1 | XP_022282599.1 | 5 - 6 | 554 | 73 | ||
| XM_022426892.1 | XP_022282600.1 | 4 - 5 | 888 | 73 | ||
| XM_022426893.1 | XP_022282601.1 | 3 - 4 | 763 | 73 | ||
More Information
Set Membership: |
|

ZNF7
Genomic Map