Assay Details
| Gene Symbol: HSD17B14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047981.1 | NP_001041446.1 | 4 - 5 | 349 | 90 | |
| XM_005615748.3 | XP_005615805.1 | 4 - 5 | 453 | 90 | ||
| XM_014114134.2 | XP_013969609.1 | 3 - 4 | 360 | 90 | ||
More Information
Set Membership: |
|

HSD17B14
Genomic Map