Assay Details
| Gene Symbol: HSD17B14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047981.1 | NP_001041446.1 | 7 - 8 | 621 | 72 | |
| XM_014114134.2 | XP_013969609.1 | 6 - 7 | 632 | 72 | ||
More Information
Set Membership: |
|

HSD17B14
Genomic Map