Assay Details
Gene Symbol: ATP6V1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628412.3 | XP_005628469.1 | 1 - 2 | 291 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
