Assay Details
| Gene Symbol: TRMT112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014121211.2 | XP_013976686.1 | 1 - 2 | 453 | 106 | |
| XM_022405749.1 | XP_022261457.1 | 1 - 2 | 447 | 106 | ||
| XM_533242.6 | XP_533242.1 | 1 - 2 | 167 | 106 | ||
More Information
Set Membership: |
|

TRMT112
Genomic Map