Assay Details
| Gene Symbol: RBKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630289.3 | XP_005630346.1 | 5 - 6 | 475 | 74 | |
| XM_014120277.2 | XP_013975752.1 | 5 - 6 | 587 | 74 | ||
| XM_014120278.2 | XP_013975753.1 | 6 - 7 | 553 | 74 | ||
| XM_022404589.1 | XP_022260297.1 | 7 - 8 | 1102 | 74 | ||
| XM_022404590.1 | XP_022260298.1 | 5 - 6 | 565 | 74 | ||
| XM_022404591.1 | XP_022260299.1 | 7 - 8 | 1111 | 74 | ||
| XM_022404592.1 | XP_022260300.1 | 6 - 7 | 544 | 74 | ||
| XM_022404593.1 | XP_022260301.1 | 6 - 7 | 713 | 74 | ||
| XM_022404594.1 | XP_022260302.1 | 5 - 6 | 616 | 74 | ||
| XM_022404595.1 | XP_022260303.1 | 7 - 8 | 582 | 74 | ||
| XM_532917.6 | XP_532917.1 | 5 - 6 | 590 | 74 | ||
More Information
Set Membership: |
|

RBKS
Genomic Map