Assay Details
| Gene Symbol: UBE2L6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_533174.4 | XP_533174.1 | 1 - 2 | 95 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_533174.4 transcripts.
Set Membership: |
|

UBE2L6
Genomic Map