Assay Details
| Gene Symbol: ATP6V1G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014376.1 | NP_001014398.1 | 2 - 3 | 179 | 83 | |
| XM_005627044.1 | XP_005627101.1 | 2 - 3 | 293 | 83 | ||
| XM_022425721.1 | XP_022281429.1 | 2 - 3 | 407 | 83 | ||
More Information
Set Membership: |
|

ATP6V1G2
Genomic Map