Assay Details
| Gene Symbol: SULT1C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_531771.5 | XP_531771.1 | 1 - 2 | 411 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SULT1C4| Gene Symbol: SULT1C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_531771.5 | XP_531771.1 | 1 - 2 | 411 | 99 | |
Set Membership: |
|