Assay Details
Gene Symbol: SMIM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627603.3 | XP_005627660.1 | 3 - 4 | 365 | 83 | |
XM_005627605.3 | XP_005627662.1 | 2 - 3 | 218 | 83 | ||
XM_022426330.1 | XP_022282038.1 | 4 - 5 | 496 | 83 | ||
XM_022426331.1 | XP_022282039.1 | 2 - 3 | 316 | 83 | ||
XM_532225.7 | XP_532225.3 | 3 - 4 | 228 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
