Assay Details
Gene Symbol: SHARPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_532352.6 | XP_532352.4 | 6 - 7 | 1313 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
