Assay Details
| Gene Symbol: RNF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629010.3 | XP_005629067.1 | 2 - 3 | 231 | 73 | |
| XM_014119683.2 | XP_013975158.1 | 2 - 3 | 240 | 73 | ||
More Information
Set Membership: |
|

RNF11
Genomic Map