Assay Details
Gene Symbol: HJV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630732.2 | XP_005630789.1 | 1 - 2 | 171 | 58 | |
XM_005630733.3 | XP_005630790.1 | 2 - 3 | 392 | 58 | ||
XM_014120583.2 | XP_013976058.1 | 2 - 3 | 398 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005630732.2, XM_005630733.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
