Assay Details
Gene Symbol: HJV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630732.2 | XP_005630789.1 | 2 - 3 | 208 | 196 | |
XM_005630733.3 | XP_005630790.1 | 3 - 4 | 429 | 149 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005630733.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
