Assay Details
| Gene Symbol: LOC474655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626440.3 | XP_005626497.1 | 2 - 3 | 1403 | 77 | |
| XM_022425197.1 | XP_022280905.1 | 3 - 4 | 1515 | 77 | ||
| XM_022425199.1 | XP_022280907.1 | 3 - 4 | 1510 | 77 | ||
More Information
Set Membership: |
|

LOC474655
Genomic Map