Assay Details
| Gene Symbol: LOC474655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626440.3 | XP_005626497.1 | 3 - 4 | 1578 | 145 | |
| XM_022425197.1 | XP_022280905.1 | 4 - 5 | 1690 | 145 | ||
| XM_022425199.1 | XP_022280907.1 | 4 - 5 | 1685 | 145 | ||
More Information
Set Membership: |
|

LOC474655
Genomic Map