Assay Details
| Gene Symbol: CKAP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630446.3 | XP_005630503.1 | 7 - 8 | 1930 | 160 | |
| XM_005630447.3 | XP_005630504.1 | 7 - 8 | 1930 | 160 | ||
| XM_022404730.1 | XP_022260438.1 | 7 - 8 | 1930 | 160 | ||
| XM_022404731.1 | XP_022260439.1 | 7 - 8 | 1930 | 160 | ||
| XM_532961.6 | XP_532961.3 | 7 - 8 | 1930 | 160 | ||
More Information
Set Membership: |
|

CKAP2L
Genomic Map