Assay Details
| Gene Symbol: CYRIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630147.3 | XP_005630204.1 | 1198 | 110 | ||
| XM_005630148.3 | XP_005630205.1 | 1188 | 110 | |||
| XM_022404468.1 | XP_022260176.1 | 1091 | 110 | |||
| XM_022404469.1 | XP_022260177.1 | 1083 | 110 | |||
| XM_022404470.1 | XP_022260178.1 | 724 | 110 | |||
More Information
Set Membership: |
|

CYRIA
Genomic Map