Assay Details
| Gene Symbol: TAMM41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206935.1 | NP_001193864.1 | 940 | 84 | ||
| XM_005632026.3 | XP_005632083.1 | 905 | 84 | |||
| XM_014121533.2 | XP_013977008.1 | 942 | 84 | |||
| XM_022406221.1 | XP_022261929.1 | 759 | 84 | |||
| XM_022406222.1 | XP_022261930.1 | 812 | 84 | |||
| XM_022406223.1 | XP_022261931.1 | 831 | 84 | |||
| XM_022406224.1 | XP_022261932.1 | 902 | 84 | |||
| XM_022406225.1 | XP_022261933.1 | 555 | 84 | |||
| XM_022406226.1 | XP_022261934.1 | 588 | 84 | |||
More Information
Set Membership: |
|

TAMM41
Genomic Map