Assay Details
| Gene Symbol: DOP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022426292.1 | XP_022282000.1 | 36 - 37 | 7094 | 72 | |
| XM_022426293.1 | XP_022282001.1 | 36 - 37 | 7067 | 72 | ||
More Information
Set Membership: |
|

DOP1A
Genomic Map