Assay Details
| Gene Symbol: MINDY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628688.3 | XP_005628745.1 | 10 - 11 | 1671 | 61 | |
| XM_005628689.3 | XP_005628746.1 | 10 - 11 | 1648 | 61 | ||
| XM_022427539.1 | XP_022283247.1 | 10 - 11 | 1671 | 61 | ||
| XM_532503.6 | XP_532503.3 | 10 - 11 | 1670 | 61 | ||
More Information
Set Membership: |
|

MINDY4
Genomic Map