Assay Details
Gene Symbol: ABRACL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_533427.6 | XP_533427.3 | 250 | 122 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_533427.6 transcripts.
Set Membership: |
![]() ![]() |
