Assay Details
| Gene Symbol: KLKB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242717.1 | NP_001229646.1 | 13 - 14 | 1734 | 82 | |
| XM_005629927.3 | XP_005629984.1 | 14 - 15 | 2457 | 82 | ||
| XM_022403603.1 | XP_022259311.1 | 14 - 15 | 2353 | 82 | ||
| GenBank mRNA | AB558589.1 | - | 13 - 14 | 1734 | 82 | |
| AB819623.1 | - | 13 - 14 | 1809 | 82 | ||
More Information
Set Membership: |
|

KLKB1
Genomic Map