Assay Details
| Gene Symbol: LYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300876.1 | NP_001287805.1 | 138 | 95 | ||
| GenBank mRNA | KJ700878.1 | - | 138 | 95 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: LYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300876.1 | NP_001287805.1 | 138 | 95 | ||
| GenBank mRNA | KJ700878.1 | - | 138 | 95 | ||
Set Membership: |
|