Assay Details
| Gene Symbol: LOC102154474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636227.3 | XP_005636284.1 | 1529 | 149 | ||
| Gene Symbol: LSM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022425855.1 | XP_022281563.1 | 4 - 5 | 366 | 149 | |
More Information
Set Membership: |
|

LOC102154474
Genomic Map