Assay Details
| Gene Symbol: COX8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252370.1 | NP_001239299.1 | 1 - 2 | 197 | 93 | |
| GenBank mRNA | DN347341.1 | - | 1 - 2 | 198 | 93 | |
More Information
Set Membership: |
|

COX8A
Genomic Map