Assay Details
| Gene Symbol: SCGB1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631626.3 | XP_005631683.1 | 1 - 2 | 137 | 80 | |
| GenBank mRNA | LC075579.1 | - | 1 - 2 | 54 | 80 | |
More Information
Set Membership: |
|

SCGB1A1
Genomic Map