Assay Details
| Gene Symbol: SCGB1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631626.3 | XP_005631683.1 | 2 - 3 | 326 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCGB1A1| Gene Symbol: SCGB1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631626.3 | XP_005631683.1 | 2 - 3 | 326 | 95 | |
Set Membership: |
|