Assay Details
| Gene Symbol: LOC479600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620502.3 | XP_005620559.1 | 2643 | 74 | ||
| XM_014113916.2 | XP_013969391.1 | 2650 | 74 | |||
| XM_014113917.2 | XP_013969392.1 | 2620 | 74 | |||
| XM_014113918.2 | XP_013969393.1 | 2604 | 74 | |||
| XM_014113919.2 | XP_013969394.1 | 2563 | 74 | |||
More Information
Set Membership: |
|

LOC479600
Genomic Map