Assay Details
| Gene Symbol: TRAPPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_537957.6 | XP_537957.2 | 2 - 3 | 272 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRAPPC2| Gene Symbol: TRAPPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_537957.6 | XP_537957.2 | 2 - 3 | 272 | 103 | |
Set Membership: |
|